missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Porimin, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin; GenBank no. AAG2164.1.
Antigen Porimin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056371 Novus Biologicals Canada
NB10056371
0.1mg Each for $255.80
NB10056371T Novus Biologicals Canada
NB10056371SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Porimin
Purified
RUO
Reacts with Human and bovine. This sequence is 100% conserved in cow, and 93% conserved in rat, mouse, and chicken.
114908
0.5mg/mL
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin; GenBank no. AAG2164.1.
Polyclonal
Primary
Western Blot
Q8N131
PBS containing 0.05% BSA