Please login to your online account to display your discounted pricing

anti-POR/Cytochrome P450 Reductase, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human CYP1B1 was used as the immunogen.
Antigen POR/Cytochrome P450 Reductase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224484

 
novus biologicals canada
NBP224484
Unlabeled 0.1mg Each for $255.80

NBP224484SS

 
novus biologicals canada
NBP224484SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human CYP1B1 was used as the immunogen.
Antigen POR/Cytochrome P450 Reductase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 84% homologous in mouse.
Gene ID (Entrez) 5447
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL