Please login to your online account to display your discounted pricing

anti-POR/Cytochrome P450 Reductase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224484SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224484SS

  • / Each
 

Description & Specifications

Specifications

Antigen POR/Cytochrome P450 Reductase
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human CYP1B1 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5447
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 84% homologous in mouse.
Target Species Human
Target Species Primate