Please login to your online account to display your discounted pricing

anti-POR/Cytochrome P450 Reductase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224484

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224484

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Format Affinity Purified
Immunogen A portion of amino acids 200-250 of human CYP1B1 was used as the immunogen.
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 5447
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 84% homologous in mouse.
Host Species Rabbit
Antigen POR/Cytochrome P450 Reductase
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO