Please login to your online account to display your discounted pricing

anti-Podocalyxin, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within an internal region (residues 350-450) of the human Podocalyxin protein. [Swiss-Prot No. O00592]
Antigen Podocalyxin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11041510

 
novus biologicals canada
NB11041510
Unlabeled 0.1mL Each for $309.05

NB11041510T

 
novus biologicals canada
NB11041510SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within an internal region (residues 350-450) of the human Podocalyxin protein. [Swiss-Prot No. O00592]
Antigen Podocalyxin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human and mouse. Immunogen sequence has 80% homology to canine and 76% homology to rat.
Gene Accession No. O00592
Gene ID (Entrez) 5420
Formulation Tris-glycine, 150mM NaCl
Concentration 1.06mg/mL