Please login to your online account to display your discounted pricing

anti-Podocalyxin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11041510SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11041510T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary
Concentration 1.06mg/mL
Gene Accession No. O00592
Monoclonal or Polyclonal Polyclonal
Test Specificity Human and mouse. Immunogen sequence has 80% homology to canine and 76% homology to rat.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide within an internal region (residues 350-450) of the human Podocalyxin protein. [Swiss-Prot No. O00592]
Regulatory Status RUO
Quantity 0.025mL
Gene ID (Entrez) 5420
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rabbit
Antigen Podocalyxin