Please login to your online account to display your discounted pricing

anti-Podocalyxin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11041510

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11041510

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human and mouse. Immunogen sequence has 80% homology to canine and 76% homology to rat.
Concentration 1.06mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen A synthetic peptide within an internal region (residues 350-450) of the human Podocalyxin protein. [Swiss-Prot No. O00592]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO
Antigen Podocalyxin
Monoclonal or Polyclonal Polyclonal
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 5420
Gene Accession No. O00592
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Format Affinity Purified