Please login to your online account to display your discounted pricing

anti-PNMA2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056634SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056634T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Gene Accession No. NP_009188.1
Gene ID (Entrez) 10687
Conjugate Unlabeled
Applications Western Blotting
Immunogen This polyclonal antibody was developed against a mixture of synthetic peptides containing amino acids 143-158 of human Ma2.
Primary or Secondary Primary
Cross Reactivity Human
Antigen PNMA2
Format Purified
Host Species Rabbit
Quantity 0.05mL
Concentration 0.5mg/mL
Regulatory Status RUO