Please login to your online account to display your discounted pricing

anti-PNMA2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056634

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056634

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_009188.1
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Regulatory Status RUO
Format Purified
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10687
Primary or Secondary Primary
Applications Western Blotting
Host Species Rabbit
Immunogen This polyclonal antibody was developed against a mixture of synthetic peptides containing amino acids 143-158 of human Ma2.
Quantity 0.2mL
Antigen PNMA2
Cross Reactivity Human