Please login to your online account to display your discounted pricing

anti-PNMA2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a mixture of synthetic peptides containing amino acids 143-158 of human Ma2.
Antigen PNMA2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056634T

 
novus biologicals canada
NB10056634SS
Unlabeled 0.05mL Each for $139.00

NB10056634

 
novus biologicals canada
NB10056634
Unlabeled 0.2mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a mixture of synthetic peptides containing amino acids 143-158 of human Ma2.
Antigen PNMA2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. NP_009188.1
Gene ID (Entrez) 10687
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL