missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PLK1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PLK1 was used as the immunogen.
Antigen PLK1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056651 Novus Biologicals Canada
NB10056651
0.2mL Each for $307.00
NB10056651T Novus Biologicals Canada
NB10056651SS
0.05mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
PLK1
Antisera
RUO
Reacts with Bovine, Dog, Human, Mouse, New World monkey, Pig, Rat and Rhesus Monkey. Predicted to react with Zebrafish, C elegans and Xenopus.
5347
A portion of amino acids 100-200 of human PLK1 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P53350
PBS containing 0.05% BSA