Please login to your online account to display your discounted pricing

anti-PLK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056651

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056651

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity Reacts with Bovine, Dog, Human, Mouse, New World monkey, Pig, Rat and Rhesus Monkey. Predicted to react with Zebrafish, C elegans and Xenopus.
Host Species Rabbit
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Quantity 0.2mL
Antigen PLK1
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P53350
Formulation PBS containing 0.05% BSA
Gene ID (Entrez) 5347
Primary or Secondary Primary
Immunogen A portion of amino acids 100-200 of human PLK1 was used as the immunogen.
Format Antiserum