missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Plexin A4, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen 1650 kMWHLVkNHEHGDQkEGDc 1667
Antigen Plexin A4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227144 Novus Biologicals Canada
NBP227144
0.1mg Each for $257.41
NBP227144SS Novus Biologicals Canada
NBP227144SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Plexin A4
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, new world monkey, panda, cow, dog, orangutan, opossum, lizard, wild turkey, zebra finch, society finch, chicken, parakeet, rhesus monkey, chimpanzee and horse, and 94% homologous in mouse, rat, zebrafish and western clawed frog.
100μg antibody in 200μL PBS containing 0.05% BSA.
1650 kMWHLVkNHEHGDQkEGDc 1667
Polyclonal
Primary
Western Blot
91584
0.5mg/mL