Please login to your online account to display your discounted pricing

anti-Plexin A4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227144SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227144SS

  • / Each
 

Description & Specifications

Specifications

Antigen Plexin A4
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen 1650 kMWHLVkNHEHGDQkEGDc 1667
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 91584
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, new world monkey, panda, cow, dog, orangutan, opossum, lizard, wild turkey, zebra finch, society finch, chicken, parakeet, rhesus monkey, chimpanzee and horse, and 94% homologous in mouse, rat, zebrafish and western clawed frog.
Target Species Human