Please login to your online account to display your discounted pricing

anti-Plexin A4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227144

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227144

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Antigen Plexin A4
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Regulatory Status RUO
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, new world monkey, panda, cow, dog, orangutan, opossum, lizard, wild turkey, zebra finch, society finch, chicken, parakeet, rhesus monkey, chimpanzee and horse, and 94% homologous in mouse, rat, zebrafish and western clawed frog.
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 91584
Concentration 0.5mg/mL
Quantity 0.1mg
Applications Western Blotting
Format Affinity Purified
Cross Reactivity Human
Immunogen 1650 kMWHLVkNHEHGDQkEGDc 1667