Please login to your online account to display your discounted pricing

anti-PKR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056399SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056399T

  • / Each
 

Description & Specifications

Specifications

Antigen PKR
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_002750.1
Host Species Rabbit
Immunogen A portion of amino acids 500-550 of mouse PRKR protein were used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5610
Test Specificity The NCBI accession number for PRKR is NP_035293. The amino acid sequence used is 92% homologous in rat. Reacts with Human. Predicted to react with Mouse and Rat.
Target Species Human