Please login to your online account to display your discounted pricing

anti-PKR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056399

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056399

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_002750.1
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Gene ID (Entrez) 5610
Cross Reactivity Human
Antigen PKR
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Purified
Applications Western Blotting
Quantity 0.1mg
Test Specificity The NCBI accession number for PRKR is NP_035293. The amino acid sequence used is 92% homologous in rat. Reacts with Human. Predicted to react with Mouse and Rat.
Concentration 0.5mg/mL
Immunogen A portion of amino acids 500-550 of mouse PRKR protein were used as the immunogen.
Regulatory Status RUO
Host Species Rabbit