Please login to your online account to display your discounted pricing

anti-PKR, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of mouse PRKR protein were used as the immunogen.
Antigen PKR
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056399

 
novus biologicals canada
NB10056399
Unlabeled 0.1mg Each for $255.80

NB10056399T

 
novus biologicals canada
NB10056399SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of mouse PRKR protein were used as the immunogen.
Antigen PKR
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The NCBI accession number for PRKR is NP_035293. The amino acid sequence used is 92% homologous in rat. Reacts with Human. Predicted to react with Mouse and Rat.
Gene Accession No. NP_002750.1
Gene ID (Entrez) 5610
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL