Please login to your online account to display your discounted pricing

anti-PKM2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the internal region of human PKM2 protein (within residues 350-450). [Swiss-Prot No. P14618]
Antigen PKM2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP148308

 
novus biologicals canada
NBP148308
Unlabeled 0.1mL Each for $329.00

NBP148308SS

 
novus biologicals canada
NBP148308SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the internal region of human PKM2 protein (within residues 350-450). [Swiss-Prot No. P14618]
Antigen PKM2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human, mouse and rat. Immunogen sequence has 95% identity to bovine.
Gene Accession No. P14618
Gene ID (Entrez) 5315
Formulation PBS, 30% glycerol
Concentration 1.41mg/mL