Please login to your online account to display your discounted pricing

anti-PKM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP148308

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP148308

  • / Each
 

Description & Specifications

Specifications

Antigen PKM2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1.41mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P14618
Host Species Rabbit
Immunogen A synthetic peptide made to the internal region of human PKM2 protein (within residues 350-450). [Swiss-Prot No. P14618]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5315
Test Specificity Human, mouse and rat. Immunogen sequence has 95% identity to bovine.
Target Species Human
Target Species Murine
Target Species Rat