Please login to your online account to display your discounted pricing

anti-PKM1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2].
Antigen PKM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP214833

 
novus biologicals canada
NBP214833
Unlabeled 0.1mL Each for $329.00

NBP214833SS

 
novus biologicals canada
NBP214833SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2].
Antigen PKM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity This antibody is specific for the M1 isoform of PKM.
Gene Accession No. P14618
Gene ID (Entrez) 5315
Formulation PBS, 30% glycerol
Concentration 1.17mg/mL