Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  BC11060477

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. BC11060477

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5314
Primary or Secondary Primary
Conjugate Unlabeled
Applications Peptide ELISA
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 100-200). [UniProt No. Q8TCZ9]
Cross Reactivity Human
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P08F94
Test Specificity Human. Immunogen sequence has 93% homology to mouse.
Concentration 0.2mg/mL
Antigen PKHD1
Host Species Rabbit
Quantity 0.1mL
Regulatory Status RUO