missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt No. Q8TCZ9]
Antigen PKHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BC11060480 Novus Biologicals Canada
BC11060480
0.1mL Each for $329.00
BC11060480T Novus Biologicals Canada
BC11060480SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
PKHD1
Affinity Purified
RUO
Human. Immunogen sequence has 86% identity to mouse.
5314
0.2mg/mL
A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt No. Q8TCZ9]
Polyclonal
Primary
Peptide ELISA
P08F94
PBS, 30% glycerol