Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3300-3400). [UniProt No. Q8TCZ9]
Antigen PKHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BC11060479

 
novus biologicals canada
BC11060479
Unlabeled 0.1mL Each for $329.00

BC11060479T

 
novus biologicals canada
BC11060479SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3300-3400). [UniProt No. Q8TCZ9]
Antigen PKHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Peptide ELISA
Test Specificity Human. Immunogen sequence has 83% identity to mouse. Customer review on mouse kidney collecting duct cell line.
Gene Accession No. P08F94
Gene ID (Entrez) 5314
Formulation PBS, 30% glycerol
Concentration 0.2mg/mL