Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 450-550). [UniProt No. Q8TCZ9]
Antigen PKHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BC11060478T

 
novus biologicals canada
BC11060478SS
Unlabeled 0.025mL Each for $109.00

BC11060478

 
novus biologicals canada
BC11060478
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 450-550). [UniProt No. Q8TCZ9]
Antigen PKHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Peptide ELISA
Test Specificity Human. Immunogen sequence has 88% identity to mouse.
Gene Accession No. P08F94
Gene ID (Entrez) 5314
Formulation PBS, 30% glycerol
Concentration 0.2mg/mL