Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  BC11060479SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. BC11060479T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. P08F94
Test Specificity Human. Immunogen sequence has 83% identity to mouse. Customer review on mouse kidney collecting duct cell line.
Concentration 0.2mg/mL
Antigen PKHD1
Format Affinity Purified
Regulatory Status RUO
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Peptide ELISA
Cross Reactivity Human
Cross Reactivity Mouse
Gene ID (Entrez) 5314
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3300-3400). [UniProt No. Q8TCZ9]
Host Species Rabbit
Quantity 0.025mL