Please login to your online account to display your discounted pricing

anti-PKHD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  BC11060478SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. BC11060478T

  • / Each
 

Description & Specifications

Specifications

Antigen PKHD1
Applications Peptide ELISA
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P08F94
Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 450-550). [UniProt No. Q8TCZ9]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5314
Test Specificity Human. Immunogen sequence has 88% identity to mouse.
Target Species Human