missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PIWIL2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 30 to 80 of mouse Mili was used as immunogen.
Antigen PIWIL2
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224590 Novus Biologicals Canada
NBP224590
0.1mg Each for $257.41
NBP224590SS Novus Biologicals Canada
NBP224590SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 30 to 80 of mouse Mili was used as immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
NP_067283.1
100μg in 200μL PBS containing 0.05% BSA
IgG
PIWIL2
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in mouse and rat, 87% homologous in human and dog, and 81% homologous in opossum.
55124
0.5mg/mL