Please login to your online account to display your discounted pricing

anti-PINK1, Polyclonal, Novus Biologicals™

$214.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human PINK1 protein (within residues 350-500). [Swiss-Prot Q9BXM7]
Antigen PINK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149678

 
novus biologicals canada
NBP149678
Unlabeled 0.1mL Each for $362.00

NBP149678SS

 
novus biologicals canada
NBP149678SS
Unlabeled 0.025mL Each for $214.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human PINK1 protein (within residues 350-500). [Swiss-Prot Q9BXM7]
Antigen PINK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Gene Accession No. Q9BXM7
Gene ID (Entrez) 65018
Formulation PBS
Concentration 1.2mg/mL