Please login to your online account to display your discounted pricing

anti-PINK1, Polyclonal, Novus Biologicals™

$164.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human PINK1 protein sequence (between residues 175-250). [Swiss-Prot Q9BXM7]
Antigen PINK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BC100494

 
novus biologicals canada
BC100494
Unlabeled 0.1mL Each for $362.00

BC100494SS

 
novus biologicals canada
BC100494SS
Unlabeled 0.025mL Each for $164.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human PINK1 protein sequence (between residues 175-250). [Swiss-Prot Q9BXM7]
Antigen PINK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Peptide ELISA
Applications Western Blot
Gene Accession No. Q9BXM7
Gene ID (Entrez) 65018
Formulation PBS
Concentration 1.0mg/mL