missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PIN1 Chicken anti-Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Manufacturer: Invitrogen PA110013
Description
Peptidyl-prolyl cis/trans isomerases , such as PIN1, catalyze the cis/trans isomerization of peptidyl-prolyl peptide bonds. PIN1 is the only PPIase that specifically binds to phosphorylated ser/thr-pro motifs to catalytically regulate the post-phosphorylation conformation of its substrates. PIN1-catalyzed conformational regulation has a profound impact on key proteins involved in the regulation of cell growth, genotoxic and other stress responses, the immune response, germ cell development, neuronal differentiation, and survival.Specifications
PIN1 | |
Polyclonal | |
PBS with 10mM sodium azide | |
Q5BIN5, Q13526, Q9QUR7 | |
0610025L01Rik, D9Bwg1161e, Peptidyl-prolyl cis/trans isomerase | |
Chicken | |
IgY | |
100 μL | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Primary | |
100055392, 100512827, 100859153, 23988, 298696, 5300, 535470 |
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Western Blot | |
Unconjugated | |
PIN1 | |
Liquid | |
PIN1 | |
Purified, recombit human Pin-1. | |
PEG precipitation | |
RUO | |
Antibody | |
Polyclonal | |
Bovine,Chicken,Equine,Human,Mouse,Porcine,Rat |