Please login to your online account to display your discounted pricing

anti-PIM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231366SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231366SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5292
Concentration 1.0mg/mL
Gene Accession No. P11309
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Host Species Rabbit
Primary or Secondary Primary
Test Specificity This antibody may detect isoform 1 as well as isoform 2 of PIM1 (Uniprot No. P11309). Human. Based upon 97% similarity to immunogen sequence, this antibody is predicted to react with Rat, Bovine and Feline (Cat). For Mouse's PIM1, the immunogen sequence shows 97% similarity to isoform 2 and 74% similarity to isoform 1.
Conjugate Unlabeled
Formulation PBS
Antigen PIM1
Format Purified
Immunogen Partial recombinant protein made to an N-terminal portion of human PIM1 (between residues 1-200) [UniProt No. P11309]
Regulatory Status RUO