Please login to your online account to display your discounted pricing

anti-PIM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231366

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231366

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 1.0mg/mL
Primary or Secondary Primary
Test Specificity This antibody may detect isoform 1 as well as isoform 2 of PIM1 (Uniprot No. P11309). Human. Based upon 97% similarity to immunogen sequence, this antibody is predicted to react with Rat, Bovine and Feline (Cat). For Mouse's PIM1, the immunogen sequence shows 97% similarity to isoform 2 and 74% similarity to isoform 1.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Formulation PBS
Gene ID (Entrez) 5292
Gene Accession No. P11309
Conjugate Unlabeled
Host Species Rabbit
Quantity 0.1mg
Cross Reactivity Human
Antigen PIM1
Format Purified
Immunogen Partial recombinant protein made to an N-terminal portion of human PIM1 (between residues 1-200) [UniProt No. P11309]