missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIG3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112547

Catalog No. 01-674-789



Description

Description

The p53 protein (393 AS, phosphoprotein) is a transcription factor, whose activity is regulated by phosphorylation. The normal function of p53 is to effect cell cycle arrest at the G1- and G2-checkpoints in response to DNA damage. The current model postulates that p53 senses DNA damage and arrests the cell cycle to allow DNA repair to take place. If repair is not successful, p53 initiates programmed cell death. In normal cells the p53 level is low. In response to DNA damage or other cellular stress signals the checkpoint function is executed by accumulation of p53.
Specifications

Specifications

PIG3
Polyclonal
PBS with 0.08% sodium azide
Q53FA7
p53 Inducible Gene 3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9540
Western Blot
Unconjugated
TP53I3
Liquid
TP53I3
Full-length recombit PIG-3 protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
SDS
Documents