Please login to your online account to display your discounted pricing

anti-PIG3 Polyclonal, Thermo Scientific™

PIG3 Polyclonal antibody PA1-12547 from Thermo Scientific™ specifically detects PIG3 in Human samples, and it is validated for Western Blot

$424.20 - $424.20

Specifications

Antigen PIG3
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Full-length recombinant PIG-3 protein.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-789

 
pierce biotechnology inc
PA112547
Unlabeled 100μg Each for $424.20
Description & Specifications

Specifications

Antigen PIG3
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Full-length recombinant PIG-3 protein.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

PIG3 Polyclonal antibody PA1-12547 from Thermo Scientific™ specifically detects PIG3 in Human samples, and it is validated for Western Blot

The p53 protein (393 AS, phosphoprotein) is a transcription factor, whose activity is regulated by phosphorylation. The normal function of p53 is to effect cell cycle arrest at the G1- and G2-checkpoints in response to DNA damage. The current model postulates that p53 senses DNA damage and arrests the cell cycle to allow DNA repair to take place. If repair is not successful, p53 initiates programmed cell death. In normal cells the p53 level is low. In response to DNA damage or other cellular stress signals the checkpoint function is executed by accumulation of p53.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Full-length recombinant PIG-3 protein.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: p53 Inducible Gene 3
Uniprot Accession: Q53FA7
Gene Symbol: TP53I3
Entrez GeneID: 9540
PA1-12547 detects PIG3 in Human samples. PA1-12547 has been successfully used in Western Blot procedures. PA1-12547 The immunogen is Full-length recombinant PIG-3 protein..