missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PIF1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611]
Antigen PIF1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP197931 Novus Biologicals Canada
NBP197931
0.1mL Each for $309.05
NBP197931SS Novus Biologicals Canada
NBP197931SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
PIF1
Affinity Purified
RUO
Q9H611
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
80119
1.16mg/mL