missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PIEZO2, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1600-1650) [UniProt Q9H5I5]
Antigen PIEZO2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178624 Novus Biologicals Canada
NBP178624
0.1mL Each for $362.00
NBP178624SS Novus Biologicals Canada
NBP178624SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
PIEZO2
Affinity Purified
RUO
Q9H5I5
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1600-1650) [UniProt Q9H5I5]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
63895
1.0mg/mL