Please login to your online account to display your discounted pricing

anti-PIEZO2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178624

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178624

  • / Each
 

Description & Specifications

Specifications

Antigen PIEZO2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q9H5I5
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1600-1650) [UniProt Q9H5I5]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 63895
Target Species Human
Target Species Murine