Please login to your online account to display your discounted pricing

anti-PICK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142829SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142829SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.14mg/mL
Isotype IgY
Formulation Borate buffered saline (BBS) and 1% BSA
Quantity 0.025mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 9463
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen PICK1
Immunogen Synthetic peptide corresponding to the C-terminus of rat PICK1 (within residues 350-416). [Swiss-Prot No. Q9EP80]
Regulatory Status RUO
Conjugate Unlabeled
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Host Species Chicken