Please login to your online account to display your discounted pricing

anti-PICK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142829

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142829

  • / Each
 

Description & Specifications

Specifications

Isotype IgY
Formulation Borate buffered saline (BBS) and 1% BSA
Concentration 0.14mg/mL
Gene ID (Entrez) 9463
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Format Affinity Purified
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Chicken
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic peptide corresponding to the C-terminus of rat PICK1 (within residues 350-416). [Swiss-Prot No. Q9EP80]
Antigen PICK1
Quantity 0.1mL