Please login to your online account to display your discounted pricing

anti-PICK1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Chicken
Isotype IgY
Immunogen Synthetic peptide corresponding to the C-terminus of rat PICK1 (within residues 350-416). [Swiss-Prot No. Q9EP80]
Antigen PICK1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142829SS

 
novus biologicals canada
NBP142829SS
Unlabeled 0.025mL Each for $109.00

NBP142829

 
novus biologicals canada
NBP142829
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Chicken
Isotype IgY
Immunogen Synthetic peptide corresponding to the C-terminus of rat PICK1 (within residues 350-416). [Swiss-Prot No. Q9EP80]
Antigen PICK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene ID (Entrez) 9463
Formulation Borate buffered saline (BBS) and 1% BSA
Concentration 0.14mg/mL