Please login to your online account to display your discounted pricing

anti-Phospho-Src pTyr527 Polyclonal, Thermo Scientific™

Phospho-Src pTyr527 Polyclonal Ab PA5-17781 from Thermo Scientific™ specifically detects Phospho-Src pTyr527 in Ck, Hu, Ms and Rt samples, and is validated for IHC(P) and WB

$448.05 - $448.05

Specifications

Antigen Phospho-Src pTyr527
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PIPA517781

 
pierce biotechnology inc
PA517781
Each for $448.05
Description & Specifications

Specifications

Antigen Phospho-Src pTyr527
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Quantity 100μL
Regulatory Status RUO
Type Primary
Formulation 100μL purified antibody in 0.01M HEPES with 100µg/ml BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Symbols SRC
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pTyr527 of human Src
Storage Requirements Store at -20°C

Phospho-Src pTyr527 Polyclonal Ab PA5-17781 from Thermo Scientific™ specifically detects Phospho-Src pTyr527 in Ck, Hu, Ms and Rt samples, and is validated for IHC(P) and WB

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene.
Host Species: Rabbit
Cross Reactivity: Chicken, Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic phosphopeptide corresponding to residues surrounding pTyr527 of human Src
Formulation: 100μL purified Antibody in 0.01M HEPES, with 100µg/ml BSA, 50% glycerol, 0.15M NaCl and no preservative; 7.5
Synonyms: OTTHUMP00000174476; OTTHUMP00000174477; OTTHUMP00000174478; proto-oncogene c-Src; proto-oncogene tyrosine-protein kinase Src; protooncogene SRC, Rous sarcoma; tyrosine kinase pp60c-src; tyrosine-protein kinase SRC-1
Uniprot Accession: P00523, P05480, P12931
Gene Symbol: SRC
Entrez GeneID: 20779, 396442, 6714, 83805
PA5-17781 detects Phospho-Src pTyr527 in Chicken, Human, Mouse and Rat samples. PA5-17781 has been successfully used in Immunohistochemistry (Paraffin) and Western Blot procedures. PA5-17781 The immunogen is Synthetic phosphopeptide corresponding to residues surrounding pTyr527 of human Src.