Please login to your online account to display your discounted pricing

anti-phospho-JNK (Thr183 and Tyr185), Polyclonal, Promega™


Rabbit Polyclonal Antibody

Manufacturer: promega corportaion  V7932

 View more versions of this product

Catalog No. PR-V7932

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Quantity 120μL
Isotype IgG
Clone Polyclonal
Primary or Secondary Primary
Antigen phospho-JNK (Thr183 and Tyr185)
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Immunogen Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme.