missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
anti-phospho-JNK (Thr183 and Tyr185), Polyclonal, Promega™
Rabbit Polyclonal Antibody
Supplier: Promega Corportaion V7931
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Many
Immunogen: Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme
Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
phospho-JNK (Thr183 and Tyr185) | |
Polyclonal | |
Rabbit | |
Affinity Purified | |
Primary |
Polyclonal | |
Unconjugated | |
Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme. | |
40 μL | |
IgG |