Please login to your online account to display your discounted pricing

anti-Phospho-JAK2 pTyr221 Polyclonal, Thermo Scientific™

Phospho-JAK2 pTyr221 Polyclonal Ab PA5-35832 from Thermo Scientific™ specifically detects Phospho-JAK2 pTyr221 in Hu, Ms and Rt samples, and it is validated for WB

$506.05 - $506.05

Products
Catalog Number Mfr. No. Price Quantity    

PIPA535832

 
pierce biotechnology inc
PA535832
Each for $506.05
Description & Specifications

Phospho-JAK2 pTyr221 Polyclonal Ab PA5-35832 from Thermo Scientific™ specifically detects Phospho-JAK2 pTyr221 in Hu, Ms and Rt samples, and it is validated for WB

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. [provided by RefSeq].
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide derived from human JAK2 around the phosphorylation site of Tyrosine 221
Formulation: 100μg purified Antibody (1mg/mL) in PBS, with 50% glycerol and 0.02% sodium azide; 7.4
Synonyms: EC 2.7.10.2, JAK-2, JAK2, Janus kinase 2, kinase Jak2
Uniprot Accession: O60674, Q62120, Q62689
Gene Symbol: JAK2
Entrez GeneID: 16452, 24514, 3717
PA5-36008 detects WWP1 in Mouse samples. PA5-36008 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-36008 The immunogen is Recombinant fragment corresponding to a region within amino acids 492 and 774 of WWP1.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.