missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phospho-Histone H3 (Ser10) Mouse anti-Human, Vertebrates, Clone: CMA312, MilliporeSigma™

Mouse Monoclonal Antibody

$127.00 - $127.00

Specifications

Clone CMA312
Host Species Murine
Isotype IgG1
Immunogen Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, phosphorylated on Ser10, conjugated to KLH.
Antigen phospho-Histone H3 (Ser10)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
05-133-6SMI View Documents Emd Millipore Corporation
051336S
10μg Each for $127.00
 
Description

Description

Anti-phospho-Histone H3 (Ser10) Antibody, clone CMA312 is a mouse monoclonal antibody for detection of phospho-Histone H3 (Ser10) also known as H3S10P, Histone H3 (phospho S10) and has been validated in western blot, ELISA, immunocytochemistry, Multiplexing and Chimmunoprecipitation.
Specifications

Specifications

CMA312
IgG1
phospho-Histone H3 (Ser10)
Purified
Chromatin Immunoprecipitation,ELISA,Immunocytochemistry,Western Blot
Purified mouse monoclonal IgG in PBS with 0.05% sodium azide.
Murine
Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, phosphorylated on Ser10, conjugated to KLH.
Monoclonal
Primary
NP_003484.1

For Research Use Only