Please login to your online account to display your discounted pricing

anti-phospho-Histone H3 (Ser10), Clone: CMA312, EMD Millipore™


Manufacturer: emd millipore corporation  051336S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-133-6SMI

  • / Each
 

Description & Specifications

Specifications

Antigen phospho-Histone H3 (Ser10)
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Immunocytochemistry
Applications Western Blot
Clone CMA312
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Vertebrate
Format Purified
Formulation Purified mouse monoclonal IgG in PBS with 0.05% sodium azide.
Gene Accession No. NP_003484.1
Gene Symbols H3.3A, H3.3B , H3F3, MGC87782, MGC87783, OTTHUMP00000035618, OTTHUMP00000035619, OTTHUMP00000035621
Host Species Murine
Immunogen Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, phosphorylated on Ser10, conjugated to KLH.
Isotype IgG1
Purification Method Protein G chromatography
Quantity 10μg
Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Vertebrate