Please login to your online account to display your discounted pricing

anti-PHD4/HIF Prolyl Hydroxylase 4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100295SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100295SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Rat
Concentration 1mg/mL
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681).
Gene Accession No. NP_808808.1
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Test Specificity Human. Does not appear to work in Mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Gene ID (Entrez) 54681
Conjugate Unlabeled
Format Affinity Purified
Regulatory Status RUO
Host Species Rabbit
Quantity 0.025mL
Antigen PHD4/HIF Prolyl Hydroxylase 4