Please login to your online account to display your discounted pricing

anti-PGP9.5 / UCHL-1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human UCHL1 protein (between residues 200-220). [UniProt No. P09936]
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300676

 
novus biologicals canada
NB300676
Unlabeled 0.1mL Each for $309.05

NB300676SS

 
novus biologicals canada
NB300676SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human UCHL1 protein (between residues 200-220). [UniProt No. P09936]
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Predicted to react with monkey, rat, pig, and horse based on 100% sequence homology.
Gene Accession No. P09936
Gene ID (Entrez) 7345
Formulation Tris-glycine, 150mM NaCl
Concentration 0.9mg/mL