Please login to your online account to display your discounted pricing

anti-PGP9.5 / UCHL-1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human UCHL1 protein (between residues 100-200). [UniProt No. P09936]
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300675

 
novus biologicals canada
NB300675
Unlabeled 0.1mL Each for $329.00

NB300675SS

 
novus biologicals canada
NB300675SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human UCHL1 protein (between residues 100-200). [UniProt No. P09936]
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Predicted to react with primate and guinea pig based on 100% sequence homology. Immunogen sequence has 99% homology to rat, pig, and horse, and 92% homology to bovine.
Gene Accession No. P09936
Gene ID (Entrez) 7345
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL